Specifications
| Lens Type | H2X |
| Certifications/Compliance | ANSI Z87.1 |
| Description | Direct/Indirect Goggle with IR3 |
| Lens Tint | IR/UV 3.0, Green |
| No. per Pack | 1 Ea. |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |