Learn More
Cytiva AcroPak™ 500 Capsules with Supor Membrane - 0.2/0.2 µm, gamma-irradiated (1/pkg)
Supplier:  Cytiva 12995
Encompass_Preferred
Fast, Final Filtration and High Throughputs for Liquid Volumes Up To 150 L•Provides higher throughputs and faster flow rates than similarly-sized competitive devices•Ideal for solutions requiring low protein binding•Saves money by increasing throughput with available built-in prefilter•100% integrity testing assures sterile filtrate•0.1 μm Supor membrane version ensures sterile, mycoplasma-free cell culture media•Fusion-welded components eliminate the potential for release of extractables from sealing adhesives•Sterilization by gamma irradiation eliminates potential contamination by EtO residuals This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0720114
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.