Learn More
ENG Scientific Capsule Stain Kit (Hiss Method)
Description
- Capsule Stain Kit
- Contents: Crystal Violet, Copper Sulfate
- Solution I - Aqueous Crystal Violet (1%) 250 mL and Solution II - Aqueous Copper Sulfate (20%) 250 mL
- For in vitro diagnostic use
- 1 Kit
- Solution made from certified dyes (when applicable)
- Recommended Procedure:
- Grow organisms in ascitic fluid or serum medium, or mix a drop of serum with specimen and from this mixture, prepare smear.
- Air dry.
- Cover smear with Sol. for 2 minutes.
- Wash off the stain with Sol. II.
- The amount of washing must be determined empirically.
- Results: Capsules appear as faint blue halos around dark blue to purple cells.
Specifications
Specifications
| Quantity | 2 x 100 mL |
| Includes | Solution I - Aqueous Crystal Violet (1%), Solution II - Aqueous Copper Sulfate (20%) |
| Product Type | Capsule Stain Kit |
| Packaging | Bottles |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.