Learn More
Erlab™ CaptairFlow 391 Clean Air Enclosure with HEPA Filter
GREENER_CHOICE
Laminar flow hood with HEPA filter and UV lights to eliminate contamination between operations.
Supplier:  Erlab™ 391LFH
Catalog No. 08-444-063
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Description
Description
Providing an ultra-clean and dust-free work area, the CaptairFlow Clean Air Enclosure is designed to prevent environmental pollution and cross-contamination. The enclosure features a high efficiency filtration system that provides a particle-free workstation around the manipulation, bringing HEPA filtered clean air down to the work surface. High efficiency UV lights are used to decontaminate the worktop from biological cross-contamination between operations. Smart Technology™ gives you an easily identifiable method of communication regarding sash status or air face velocity: check sash, pre-filter, HEPA/ULPA, and fan status via an LED band of light around the enclosure called a Smart-Light.
  • For non-pathogenic cell cultures, in-vitro cultures, microbiology, homeopathic preparations, electronics, and optics
  • Smart Technology™ tool consists of Smart-Light Communication, chemical sensors, real-time status, and eGuard app
  • Comes equipped with needed quantity of GF4.HP filter and Pre-Filter M for first time use
  • Units do not come standard with a stainless steel work surface
Specifications
Specifications
Flow Hood
55 W
241.3 mm
99.995%
Protecting Samples In Clean Bench Procedures
LED, UV
9.5 in.
1079 mm
24.25 in.
170 lb.
90/240 V
55 dBA
28 in.
H14 HEPA
GF4.HP, Pre-filter M, and Pre-filter XLS
Corrosion Resistant Electro-galvanized Steel Coated with Anti-acid Polymer and Polypropylene
711 mm
39.75 in.
50/60 Hz
616 mm
77.1 kg
0.45 A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.