Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Capto™ L Chromatography Medium

Catalog No. p-4519674 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
200 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-603 5 mL
45-002-601 25 mL
09-928-076 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-002-603 Supplier Cytiva Supplier No. 17547806
Only null left
Add to Cart
Add to Cart

Capto L is a BioProcess chromatography medium (resin) for the capture of antibodies and antibody fragments.

Capto L is suitable for the capture of a wide range of antibody fragments such as Fabs, single-chain variable fragments (scFv), and domain antibodies (Dabs).

  • High binding capacity, low ligand leakage, and selectivity for a broad range of antibody fragments reduces process time and amount of medium.
  • Enables a platform approach for purification of antibody fragments.
  • Increased productivity, process economy, and process design flexibility due to rigid matrix.
  • The high binding capacity and selectivity for a broad range of antibody fragments makes Capto L an excellent capture platform for antibody fragments containing kappa light chains.
  • Using Capto L requires minimal optimization and it produces high purity in one separation step.
  • Capto L provides several advantages in manufacturing processes because the high selectivity of affinity capture reduces the number of subsequent polishing steps.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Capto L chromato graphy media
Quantity 5 mL
Height (English) 7.87 in.
Height (Metric) 200mm
Flow Rate <500 cm/hr.
Ligand Type Recombinant Protein L (E. coli), mammalian free
pH Range 2 to 10
Particle Size 85μm
Support Type Highly Cross-linked Agarose
Content And Storage 2° to 8°C
For Use With (Application) High binding capacity, low ligand leakage and selectivity for a broad range of Antibody fragments reduces process time and amount of medium
Matrix Affinity Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.