Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Capto S™ ImpAct Chromatography Media

Catalog No. p-4917497 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-003-200 25 mL
45-003-201 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-003-200 Supplier Cytiva Supplier No. 17371701
Only null left
Add to Cart
Add to Cart

Capto S ImpAct is a strong cation exchange chromatography medium (resin) developed for polishing of monoclonal antibodies (MAbs) and other biomolecules.

  • High binding capacity, > 100 mg MAb/mL medium.
  • Efficient aggregate removal at high load of monoclonal antibodies.
  • High-resolution polishing based on the well-established Capto platform.
  • Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
  • High productivity enabling cost-efficient manufacturing.
  • Security of supply and comprehensive regulatory support.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Capto S ImpAct Chromatography Media
Quantity 25 mL
Material High-flow agarose
pH Range 4 to 12
Particle Size 50μm
Matrix Ion Exchange Chromatography Media
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.