Learn More
Cayman Chemical CaptoprIl-d3 250ug
SDP
Supplier:  Cayman Chemical 25235250 UG
Encompass_Preferred
An antiplatelet drug that inhibits both TXA2 receptors (Kd 325 nM) and TXA2 synthase significantly inhibits basal and LPS-stimulated TXA2 synthesis by rat peritoneal macrophages when used at 0.5 mM
Catalog No. 50-293-2632
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.