Learn More
Labconco™ Capture™ BT Fuming System Accessory, Casing Holder Kit
GSA_VASDP
Catalog No. 3181700 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
CApture BT Fuming System
Catalog No. For Use With (Equipment)
31-817-00 CApture BT Fuming System
1 options
Catalog No. 31-817-00 Supplier Labconco™ Supplier No. 3181700
Add to Cart
Add to Cart
For use with CApture BT Fuming System
  • Stainless steel housing provides 45 numbered standoffs to hold shell casings during fuming
  • Minimum 0.22 caliber (upside down placement)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Fuming System Chamber
Material Stainless Steel
Dimensions (L x W x H) 12.7 x 12.7 x 63.5 cm (5 x 5 x 25 in.)
For Use With (Equipment) CApture BT Fuming System
Weight (English) 10 lb.
Weight (Metric) 5 kg