Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ AAT-XL Affinity Matrix
GREENER_CHOICE

Catalog No. 2943832005
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
250 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-438-32005 5 mL
29-438-32010 10 mL
29-438-32050 50 mL
19-438-32250 250 mL
19-438-3201L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 29-438-32005 Supplier Thermo Scientific™ Supplier No. 2943832005
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ AAT-XL Affinity Matrix has been specifically designed for the purification of native human alpha-1 antitrypsin (AAT) from complex sources like plasma/serum and recombinant human AAT from cell culture supernatants.

CaptureSelect AAT-XL Affinity Matrix has been specifically designed for the purification of native human alpha-1 antitrypsin (AAT) from complex sources like plasma/serum and recombinant human AAT from cell culture supernatants. This affinity matrix offers mild elution conditions at pH 6. The high selectivity and yield obtained using CaptureSelect AAT-XL Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in a single step.

Features of this affinity matrix include:

  • Highly selective for native and recombinant human AAT
  • High recovery and yield obtained in a single purification step
  • Mild elution at pH 6
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect AAT-XL affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: 15 mg AAT/mL resin
  • Elution conditions: 20 mM Tris, 2.0 M MgCl2, pH 6.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity 15 mg AAT/mL resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based, epoxide-activated
Particle Size 65 ±10 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Ligand Type AAT-XL affinity ligand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.