Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ C-tagXL MiniChrom™ Pre-packed Column
GREENER_CHOICE

Catalog No. 5943072001
Change view
Click to view available options
Column Capacity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Column Capacity
59-430-72001 1 mL
59-430-72005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 59-430-72001 Supplier Thermo Scientific™ Supplier No. 5943072001
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ C-tagXL MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

CaptureSelect C-tagXL MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

These pre-packed columns are ready to connect and have the following features:

  • Dimensions of 1 mL (0.8 x 2 cm) and 5 mL (0.8 x 10 cm)
  • Direct connection to standard chromatography systems
  • Packed with CaptureSelect C-tagXL Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates

The 1-mL columns can be used for quick screening of application feasibility and lab-scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5-mL column allows initial process development on a bench scale.

CaptureSelect C-tagXL Affinity Matrix has a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine). CaptureSelect C-tagXL Affinity Matrix purifies the C-terminal tagged proteins with high affinity and selectivity, even in the presence of urea and guanidine HCl, from complex mixtures like cell culture harvests and periplasmic fractions in a one-step process. Mild elution conditions at neutral pH can be applied using magnesium chloride or propylene glycol, which ensures high-activity recoveries of pH-sensitive target proteins. The affinity resin recognizes the E-P-E-A tag sequence when fused either directly to the C-terminus of a protein or through a linker between the C-terminus and the E-P-E-A tag.

Features of this affinity matrix include:

  • Mild elution, making it suitable for pH-sensitive proteins
  • Binding of the tagged proteins under denaturing conditions (like dissolved inclusion bodies)
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect C-tagXL affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: 400 nmol/mL resin depending on flow rate, column height, contact time
  • Elution conditions: Acidic: 20 mM citric acid or acetic acid, pH 3–4; 100 mM glycine buffer, pH 2–3. Neutral (pH 7.0–7.4): 20 mM Tris, 2.0 M MgCl2; 20 mM Tris, 1.0 M NaCl, 50% (v/v) propylene glycol; 20 mM Tris, 2 mM 'S-E-P-E-A' peptide
  • Column dimensions: 1 mL (0.8 x 2 cm) and 5 mL (0.8 x 10 cm)
  • Flow characteristics: 75–150 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol

Specifications

Flow Rate 75 to 150 cm/hr. (Up to 2 bar)
Stationary Phase C-tagXL
Column Capacity 1 mL
Length (Metric) 2 cm
Quantity 1 mL
Particle Size 65 ±10 μm
Type Column
Diameter (Metric) Inner 0.8 cm
Content And Storage Store in refrigerator (2°C to 8°C).
Binding Capacity 400 nmol/mL
Product Line CaptureSelect
Ligand Type C-tagXL affinity ligand
Matrix Agarose-based, epoxide-activated
Shipping Condition Wet Ice
Column Format MiniChrom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.