Learn More
Thermo Scientific™ CaptureSelect™ C-tagXL Pre-packed Column

Description
CaptureSelect C-tagXL Affinity Matrix is an improved version of CaptureSelect C-tag Affinity Matrix with a significantly increased binding capacity, offered here in pre-packed format.
Like C-tag Affinity Matrix, C-tagXL Affinity Matrix combines a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine) with the benefits of a robust and high quality affinity matrix. CaptureSelect C-tagXL Affinity Matrix purifies C-terminal tagged proteins with high affinity and selectivity, even in the presence of urea and guanidine HCl, from complex mixtures like cell culture harvests and periplasmatic fractions in a one-step process. Mild elution conditions at neutral pH can be applied using magnesium chloride or propylene glycol, which ensures high activity recoveries of pH-sensitive target proteins. The affinity resin recognizes the E-P-E-A tag sequence when fused either directly to the C-terminus of a protein or through a linker between the C-terminus and the E-P-E-A tag.
The affinity matrix is offered in a pre-packed format as a set of 5 x 1 mL columns, or as one 5 mL column, with connectors that are compatible with standard chromatography equipment.
Specifications
Specifications
| Content And Storage | Store in refrigerator (2°C to 8°C). |
| Shipping Condition | Room Temperature |
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Quantity | 1 x 5 mL Column |
| Type | Pre-packed Column |
| Stationary Phase | Anti-C-tagXL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.