Learn More
Thermo Scientific™ CaptureSelect™ Factor X Affinity Matrix

Description
CaptureSelect Factor X Affinity Matrix was specifically designed for the purification of human Factor X from plasma and recombinant sources.
Features of the CaptureSelect Factor X Affinity Matrix include:
- Highly selective for human Factor X with no affinity for other human plasma proteins
- Binds both Factor X and Factor Xa in the presence of calcium
- Enables efficient purification of Factor X with high yields and retained activity
- Mild elution at neutral pH using EDTA
- Offers excellent scalability
- Non-animal-derived
Coagulation Factor X is a vitamin K-dependent, liver-produced serine protease that serves as the first enzyme in the coagulation cascade to form fibrin. It is a two-chain glycoprotein with a molecular weight of approximately 59 kDa. Factor X normally circulates in the plasma as an inactive molecule with activation of Factor X involved in both the intrinsic and extrinsic coagulation pathways.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics
- Matrix: agarose-based
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect Factor X affinity ligand
- Ligand coupling method: epoxide
- Binding capacity: >10 mg/mL resin
- Binding conditions: 50 mM Tris, 150 mM NaCl, 5 mM CaCl2, pH 7
- Elution conditions: 50 mM Tris, 150 mM NaCl, 5 mM EDTA, pH 7
- Flow characteristics: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20% (v/v) ethanol
Specifications
Specifications
| Binding Capacity | >10 mg/mL |
| Flow Rate | 50 to 200 cm/hr. (Up to 2 bar) |
| Matrix | Agarose-based |
| Particle Size | 65 ±10 μm |
| Form | Resin Slurry |
| Product Line | CaptureSelect |
| Quantity | 250 mL |
| Type | Affinity Matrix |
| Ligand Type | Factor X affinity ligand |
| Stationary Phase | Factor X |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.