Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ FcXP Affinity Matrix
GREENER_CHOICE

Catalog No. 1943712005
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
250 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-437-12005 5 mL
19-437-12010 10 mL
19-437-12050 50 mL
19-437-12250 250 mL
19-437-1201L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-437-12005 Supplier Thermo Scientific™ Supplier No. 1943712005
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ FcXP Affinity Matrix is the next-generation CaptureSelect FcXL matrix, specifically designed for higher binding capacity.

CaptureSelect FcXP Affinity Matrix is the next-generation CaptureSelect FcXL matrix, specifically designed for higher binding capacity. The selectivity remains identical; the ligand recognizes the CH3 domain of all human IgG subclasses (IgG1, 2, 3, and 4), while maintaining the elution characteristics of CaptureSelect FcXL Affinity Matrix, but the binding capacity is increased (>40 g IgG/L).

Features of this affinity matrix include:

  • Mild elution, making it suitable for Fc fusion proteins
  • Human-specific, no binding to bovine antibodies
  • Excellent scalability
  • Non-animal derived

CaptureSelect FcXP Affinity Matrix can be used for the purification of IgG and Fc fusion proteins directly from complex source materials and enables a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect FcXP affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: 40–50 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 20 mM citric acid or acetic acid, pH 3–4, or 100 mM Tris, 2.0 M MgCl2, 40% (v/v) propylene glycol, pH 7
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity 40 to 50 g IgG/L resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based, epoxide activated
Particle Size 65 ±10 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Ligand Type FcXP affinity ligand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.