Learn More
Thermo Scientific™ CaptureSelect™ Human Albumin Affinity Matrix

Description
CaptureSelect Human Albumin Affinity Matrix has been specifically designed for the purification of human albumin and albumin fusion proteins.
Features of this affinity matrix include:
- Highly selective for human albumin
- Unique elution characteristics allowing for gentle elution of albumin fusion proteins
- Excellent scalability
- Non-animal-derived
The high selectivity and yields obtained using CaptureSelect Human Albumin Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step. The mild elution conditions make the resin suitable for the purification of albumin fusion proteins, especially those that are pH sensitive.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main Characteristics:
- Matrix: agarose-based, aldehyde-activated
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect HSA affinity ligand
- Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
- Binding capacity: ∼15 mg albumin/mL resin
- Elution conditions (neutral pH): 20 mM Tris, pH 7.4 with 2 M MgCl2 or with 1 M NaCl + 50%(v/v) propylene glycol or with 1 M NaCl + 0.5 M L-arginine
- Elution conditions (acidic pH): 0.1 M glycine, pH 3.0
- Flow characteristics: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20%(v/v) ethanol
Specifications
Specifications
| Binding Capacity | ∼15 mg albumin/mL resin |
| Flow Rate | 50 to 200 cm/hr. (Up to 2 bar) |
| Matrix | Agarose-based, aldehyde-activated |
| Content And Storage | Store at 2°C to 8°C. |
| Particle Size | 65 ±10 μm |
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Quantity | 10 mL |
| Type | Affinity Matrix |
| Ligand Type | HSA affinity ligand |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.