Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ Human Fab-kappa Kinetics Biotin Conjugate
GREENER_CHOICE

Catalog No. 7103302100
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-033-02100 100 μg
71-033-02500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-033-02100 Supplier Thermo Scientific™ Supplier No. 7103302100
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ Human Fab-kappa Kinetics Biotin Conjugate is a bi-functional ligand construct consisting of 2 different 13 kDa recombinant single domain antibody fragments (affinity ligands).

CaptureSelect Human Fab-kappa Kinetics Biotin Conjugate is a bi-functional ligand construct consisting of 2 different 13 kDa recombinant single domain antibody fragments (affinity ligands).

The CaptureSelect anti-LC-kappa (Hu) ligand that specifically binds to the constant domain of human kappa light chains is genetically fused to the CaptureSelect anti-IgG-CH1 ligand that specifically binds to the CH1 domain of all four human IgG subclasses. The bi-functional affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Human Fab-kappa Kinetics Biotin Conjugate allows you to:

  • Screen Fab-antigen interactions—the Human Fab-kappa Kinetics Biotin Conjugate allows high affinity capture of human IgG Fab fragments that possess a kappa light chain. Extreme low off-rates [k(diss)] are achieved through avidity by targeting the CL-kappa domain of the Fab (light chain) and its IgG CH1 domain (heavy chain) simultaneously, thereby enabling Fab screening on target binding kinetics.

Applications for CaptureSelect Human Fab-kappa Kinetics Biotin Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Form Liquid
Product Line CaptureSelect
Quantity 100 μg
Type Conjugate
Label or Dye Biotin & Other Haptens
Ligand Type Antibody
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.