Learn More
Thermo Scientific™ CaptureSelect™ IgG3 (Hu) Affinity Matrix
Catalog No. 191304005 Shop All MSA Products
Change view
Click to view available options
5 mL
Catalog No.
19-130-4005 5 mL
1 options
Catalog No. 19-130-4005 Supplier Thermo Scientific™ Supplier No. 10063763
CaptureSelect™ IgG3 (Hu) Affinity Matrix can be used for the purification and isolation of IgG3, without cross-reactivity to other immunoglobulins.
CaptureSelect™ IgG3 (Hu) Affinity Matrix can be used for the purification and isolation of IgG3, without cross-reactivity to other immunoglobulins, like other human IgG subclasses, IgA, and IgM, from complex sources like plasma, serum, and cell culture supernatants.