Learn More
Thermo Scientific™ CaptureSelect™ TSH Affinity Matrix

Description
CaptureSelect TSH Affinity Matrix was specifically designed for the purification of thyroid-stimulating hormone (TSH) from recombinant sources.
Some features of CaptureSelect TSH Affinity Matrix include:
- Highly selective for thyroid-stimulating hormone with no affinity for the other gonadotropins (FSH, LH, and HCG)
- Enables efficient purification of recombinant TSH with high yields and retained activity
- Offers excellent scalability
- Non–animal-derived
The high selectivity and yields obtained using CaptureSelect TSH Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics:
- Matrix: agarose bead
- Average particle size: 65 μm
- Ligand: CaptureSelect TSH affinity ligand
- Ligand coupling method: epoxide
- Binding capacity: >5 mg/mL of resin
- Elution conditions: 0.1 M citrate buffer, pH 3.0
- Flow characteristics: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20% (v/v) ethanol
Specifications
Specifications
| Binding Capacity | >5 mg/mL |
| Flow Rate | 50 to 200 cm/hr. (Up to 2 bar) |
| Matrix | Agarose-based |
| Content And Storage | Store in refrigerator (2°C to 8°C). |
| Shipping Condition | Wet Ice |
| Particle Size | 65 μm |
| Form | Agarose Bead |
| Product Line | CaptureSelect |
| Quantity | 1 L |
| Type | Affinity Matrix |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.