Learn More
Invitrogen™ CAR Polyclonal Antibody

Description
PA1-9541 detects CXADR in human samples. PA1-9541 has been successfully used in Western blot procedures. By Western blot, PA1-9541 detects a 38 kDa protein representing CXADR. PA1-9541 immunogen is a purified recombinant CXADR corresponding to the chromosome 21 gene (Human).
Specifications
Specifications
| Antigen | CAR |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CXADR |
| Gene Accession No. | P78310 |
| Gene Alias | 2610206D03Rik; 46 kD coxsackievirus and adenovirus receptor (CAR) protein; AU016810; AW553441; CAR; Car1; CAR4/6; coxsackie virus and adenovirus receptor; coxsackievirus and adenovirus receptor; Coxsackievirus and adenovirus receptor homolog; coxsackievirus B-adenovirus receptor; CVB3-binding protein; Cxadr; HCAR; HCVADR; MCAR; MCVADR; rCAR |
| Gene Symbols | CXADR |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.