Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbazeran citrate, Tocris Bioscience™

Catalog No. 442010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-201-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-201-0 Supplier R&D Systems Supplier No. 4420/10
This item is not returnable. View return policy

Aldehyde oxidase substrate; PDE inhibitor

Aldehyde oxidase (AO) substrate. Exhibits activities as a phosphodiesterase inhibitor; produces concentration-dependent positive inotropic responses in isolated right atria.

Chemical Identifiers

CAS 153473-94-0
Molecular Formula C24H32N4O11
Molecular Weight (g/mol) 552.537
InChI Key QKMRPDVEKBYWPF-UHFFFAOYSA-N
Synonym carbazeran citrate, ethyl 1-6,7-dimethoxy-1-phthalazinyl-4-piperidinyl carbamate citrate
PubChem CID 90488940
IUPAC Name [1-(6,7-dimethoxyphthalazin-1-yl)piperidin-4-yl] N-ethylcarbamate;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCNC(=O)OC1CCN(CC1)C2=NN=CC3=CC(=C(C=C32)OC)OC.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 552.53
Percent Purity >98%
Chemical Name or Material Carbazeran citrate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.