Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbazole 97.0+%, TCI America™
SDP

Catalog No. C0032100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0032100G 100 g
C003225G 25 g
C0032500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0032100G Supplier TCI America Supplier No. C0032100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86-74-8
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004960
InChI Key UJOBWOGCFQCDNV-UHFFFAOYSA-N
Synonym carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn
PubChem CID 6854
ChEBI CHEBI:27543
IUPAC Name 9H-carbazole
SMILES N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 245°C
Color White-Yellow
Boiling Point 355°C
Quantity 100 g
UN Number 3077
Formula Weight 167.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Carbazole
EINECSNumber (5)-0150
RTECSNumber FE3150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.