Learn More
Enzo Life Sciences Carbenicillin . disodium salt (250mg). CAS: 4800-94-6
Supplier: Enzo Life Sciences ALX380270M250
Analog to ampicillin (ALX-380-268). Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-positive and Gram-negative bacteria and Pseudomonas. Alternative name: alpha-Carboxybenzylpenicillin . 2Na. Purity: ≥92% (Assay). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.