Learn More
Growcells CARBENICILLIN DISODIUMSALT10GM
Supplier:  Growcells MBPC2202
Encompass_Preferred
Carbenicillin Disodium Salt is a penicillin derivative that interferes with cell wall synthesis in gram negative and positive bacteria used for ampicillin-resistant plasmids. Carbenicillin maintains stability after being autoclaved.
Catalog No. 50-103-4870
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.