Learn More
Apexbio Technology LLC Carbetocin 37025-55-1 10mg
SDP
Supplier:  Apexbio Technology LLC B573610
Encompass_Preferred
Carbetocin (CAS 37025-55-1) is a synthetic peptide analogue of oxytocin designed to bind and activate oxytocin receptors on uterine smooth muscle By stimulating these receptors carbetocin induces uterine contractions thereby facilitating myometrial contraction and reducing postpartum uterine bleeding Its pharmacological profile includes prolonged uterotonic activity compared to native oxytocin Carbetocin is employed in biomedical research to study uterine contractility receptor agonism and the regulation of postpartum hemorrhage providing a model compound for investigations involving peptide hormone analogues in reproductive biology
Catalog No. 50-000-88529
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.