Learn More
Invitrogen™ Carbofuran Monoclonal Antibody (3H2)

Description
Antibody tested in indirect ELISA.
Specifications
Specifications
| Antigen | Carbofuran |
| Applications | ELISA, Immunohistochemistry, Lateral Flow, SDS-Page |
| Classification | Monoclonal |
| Clone | 3H2 |
| Concentration | 3.85 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | Carbofuran; Furadan |
| Host Species | Mouse |
| Immunogen | Conjugate of CAR hapten and KLH. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.