Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbofuran, SPEX CertiPrep™
SDP

Catalog No. S740 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S-740 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S-740 Supplier SPEX CertiPrep Supplier No. S740
Add to Cart
Add to Cart

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Carbofuran1563-66-21000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 1563-66-2 , 67-56-1
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.256
InChI Key DUEPRVBVGDRKAG-UHFFFAOYSA-N
PubChem CID 2566
ChEBI CHEBI:34611
IUPAC Name (2,2-dimethyl-3H-1-benzofuran-7-yl) N-methylcarbamate
SMILES CC1(CC2=C(O1)C(=CC=C2)OC(=O)NC)C

Specifications

Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol
Quantity 1 mL
Chemical Name or Material Carbofuran
Hazard Category Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness.
DOTInformation Nonhazardous
Recommended Storage Ambient
WARNING: Cancer - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.