Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Carbol Fuchsin Solution, 0.3%
SCI_ED

Catalog No. S25233 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25233 100 mL
S25233A 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25233

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Carbol Fuchsin Solution, 0.3%
CAS 632-99-5, 64-17-5, 108-95-2, 7732-18-5
Assay Percent Range <0.3%
Synonym Ziehl-Neelsen
Packaging 0.30%
Color Red
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.