Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbomer 934P, Resin, NF, Spectrum™ Chemical
SDP

Catalog No. 18601452 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
22 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-452 2.5 kg Poly Pail
18-601-450 100 g Poly Bottle
18-601-451 500 g Poly Pail
18-601-453 22 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-452 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA1842.5KG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

CA184, 9003-01-4

Spectrum™ Chemical Carbomer 934P, Resin, NF polymers can provide bioadhesion, taste-masking and good binding characteristics that provide highly effective controlled-release properties. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9003-01-4

Specifications

CAS Max % 100%
Quantity 2.5 kg
Grade NF
Loss on Drying 2%
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.