Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ CARBON ARMOUR™ FR Base Layer Shirt
SDP

Catalog No. 17355092 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-092 Large
17-355-090 Small
17-355-091 Medium
17-355-093 X-Large
17-355-094 2X-Large
17-355-095 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-355-092 Supplier National Safety Apparel™ Supplier No. BSTAVLSLG
Add to Cart
Add to Cart
This item is not returnable. View return policy

FR base layer shirts are constructed of exceptionally soft, breathable, and lightweight fabric.

  • Inherently flame resistant, protection will not wash or wear out
  • Perfect for layering under Primary PPE for additional FR and heat protection
  • Exceptionally soft, breathable, and lightweight
  • Moisture wicking and quick drying for superior comfort in high heat settings
  • Rib knit cuff bottoms
  • Front fly
  • Made in the USA

Specifications

Size Large
Gender Unisex
Material Polyester, Carbon
Color Gray
Neck Style Collar
Sleeve Style Long Sleeve
Closure Type Pull On
Disposable Reusable
Product Type Shirt
Resistance CAT 2
Hazard Risk Category CAT 2
Arc Flash Rating 12 cal/cm²
Certifications/Compliance NFPA 70 E, ASTM F2700, CSA Z462, OSHA 1910.269, ASTM F1506
Style Base Layer Shirt
For Use With (Application) Molten Metals
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.