Learn More
Cole-Parmer CARBON DIGTIL CALIPER 0-4IN
Supplier:  Cole-Parmer EW9715216
Encompass
Digital Caliper with Calibration Carbon Fiber 0-4in. Includes NIST traceable calibration certificate with data.Measure inside / outside / step / adjustable zero inch/metric conversion.Designed with 0-4 inch range with 0.001inch resolution and plus/minus 0.01inch accuracy.
Catalog No. NC3825448
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.