Learn More
MilliporeSigma™ Supelco™ Carbon Dioxide Trap, Fitting size 1/4 in.
Catalog No. 111016412 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
250 cc
Catalog No. Volume (Metric)
11-101-6412 250 cc
1 options
Catalog No. 11-101-6412 Supplier MilliporeSigma™ Supelco™ Supplier No. 503215
Constructed of borosilicate glass, and the fittings are nickel-plated and have sintered stainless steel frits
Sodium hydroxide nonfibrous silicate material is universally accepted for its high absorptive capacity and indicating properties for carbon dioxide. Typically, this material will absorb 20-30% of its weight in carbon dioxide before replacement of the saturated material is required. As carbon dioxide is absorbed, the greenish-brown material turns white because of the formation of sodium carbonate. The trap body is constructed of borosilicate glass, and the fittings are nickel-plated and have sintered stainless steel frits. A moisture trap should be installed downstream from this unit to absorb the moisture that is produced. Note: Exercise extreme caution when refilling the trap due to the caustic nature of the absorbent. Dimensions: 100 cc Traps: 123/4 in. (32.4 cm) long x 13/4 in. (44 mm). Diameter: 250 cc Traps: 171/2 in. (44.4 cm) long x 2 in. (51 mm) diameter
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 250 cc