Learn More
Sigma Aldrich Fine Chemicals Biosciences Carbon nanotube sngle wal250MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 652490250MG
Encompass
Single walled nanotubes were prepared by the electric arc discharge method. The SWNTs were functionalized by purifying them in nitric acid. The carboxylic acid (-COOH) functionality along the length of the nanotube can be derivatized with a variety of functional groups. The average diameter of the nanotubes is 1.4nm /- 0.1nm. Transport properties of the SWNTs in soil was studied.
Catalog No. 50-290-7400
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.