Learn More
Eisco™ Carbon Paper Disc
SCI_ED
Catalog No. S12952 Shop All Eisco Products
Change view
Click to view available options
Quantity:
100/Box
Catalog No. Quantity
S12952 100/Box
1 options
This item is not returnable. View return policy
Catalog No. S12952 Supplier Eisco™ Supplier No. PH0353C
Add to Cart
Add to Cart
This item is not returnable. View return policy
Supplied as 100 discs per box.
  • Made from paper
  • Box of 100
Specifications
Product Type Carbon Paper Disc
Description 100 discs per box
Material Carbon Paper
For Use With (Equipment) For use with Ticker timers ( PH0353A)
Quantity 100/Box
Diameter (English) 2 in.
Diameter (Metric) 50 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov