Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Carbon / Technora Full Face Hood

Catalog No. 19100457 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Design Type:
Straight Cut Design
Tri-Cut Design
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Design Type
19-100-457 Straight Cut Design
19-100-458 Tri-Cut Design
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-100-457 Supplier PIP™ Supplier No. 9068416CT
Add to Cart
Add to Cart
This item is not returnable. View return policy

PIP™ Carbon / Technora Full Face Hood features lightweight carbon fiber for excellent fire resistance

  • Carbon fiber is lightweight and provides excellent fire resistance
  • Technora™ adds strength, durability, and helps hood retain its shape when launder
  • Full face coverage
  • TPP Rating 28
  • Meets NFPA 1971 Requirements
  • One Size Fits Most

Specifications

Product Type Fire Protectant Hood
Size Universal
Material Nomex
Color Black
Closure Type Pull-over
Disposable Reusable
Design Type Straight Cut Design
Quantity 1 Ea.
Includes Hood Only
For Use With (Application) Heat/Fire Applications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.