Learn More
PIP™ Carbon / Technora Full Face Hood
Catalog No. 19100458 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Design Type:
Straight Cut Design
Tri-Cut Design
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Design Type
19-100-458 Tri-Cut Design
19-100-457 Straight Cut Design
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-100-458 Supplier PIP™ Supplier No. 9068416CBX
  • $137.00
    $109.65 / Each of 1
    Save $27.35
    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
PIP™ Carbon / Technora Full Face Hood features lightweight carbon fiber for excellent fire resistance
  • Carbon fiber is lightweight and provides excellent fire resistance
  • Technora™ adds strength, durability, and helps hood retain its shape when launder
  • Full face coverage
  • TPP Rating 28
  • Meets NFPA 1971 Requirements
  • One Size Fits Most
Specifications
Product Type Fire Protectant Hood
Size Universal
Material Nomex
Closure Type Pull-over
Disposable Reusable
Design Type Tri-Cut Design
Quantity 1 Ea.
Includes Hood Only
For Use With (Application) Heat/Fire Applications