Learn More
Boston Bioproducts Inc Carbonate-Bicarbonate Buffer (1 M, pH 9.0) - 250ml
Supplier:  Boston Bioproducts Inc BB64250ML
Encompass_Preferred
The Carbonate-Bicarbonate Buffer is commonly used to maintain pH levels above physiological pH, and is used extensively in molecular and cell biology. Applications of this buffer include  its use as a coating buffer for microtiter plates prior to enzyme-linked immunosorbent assay (ELISA) or sandwich ELISA, and for protein and antibody conjugation procedures. The bicarbonate buffering system is also important in many different cellular processes such as to maintain pH.
Catalog No. NC2681858
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.