Learn More
Description
Specifications
Specifications
| Assay Range | 31.25 pg/mL |
| Assay Sensitivity | 31.25-2000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Line | Novus |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Carbonic Anhydrase VIII/CA8. |
| Target Species | Mouse |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
