Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Carbonic Anhydrase XIII Recombinant Rabbit Monoclonal Antibody (103), Invitrogen™

Catalog No. p-7240532
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543470 50 μL
PIMA543063 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543470 Supplier Invitrogen™ Supplier No. MA543470
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human Carbonic Anhydrase XIII/CA13

Carbonic anhydrase XII (CA XII), encoded by the CA12 gene, is one of the enzyme isoforms in the carbonic anhydrase family that catalyzes the reversible hydration of carbon dioxide into bicarbonate and protons. CA XII is a transmembrane protein expressed in various tissues, including the kidneys, intestines, and lungs, playing crucial roles in maintaining acid-base balance, pH regulation, and electrolyte transport. Its activity is significant in organs where fluid secretion and absorption are essential processes, impacting functions such as respiration and renal bicarbonate reabsorption. CA XII is involved in physiological processes like cell proliferation and differentiation by modulating the extracellular and intracellular pH, influencing tumor microenvironment dynamics. Aberrant expression of CA XII has been noted in several cancers, including breast and colorectal cancers, where it supports tumor growth and metastasis by maintaining the acid-base homeostasis conducive to cancer cell survival. Due to its surface expression and role in maintaining pH balance, CA XII is a target of interest for therapeutic interventions aiming to disrupt tumor progression and enhance the efficacy of cancer treatments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase XIII
Applications ELISA
Classification Recombinant Monoclonal
Clone 103
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ca13
Gene Accession No. Q8N1Q1
Gene Alias 2310075C21Rik; Ca13; Car13; Carbonate dehydratase XIII; carbonic anhydrase 13; Carbonic anhydrase XIII; CAXIII; CA-XIII; RGD1560453
Gene Symbols Ca13
Host Species Rabbit
Immunogen Recombinant Human Carbonic Anhydrase XIII/CA13 protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 377677
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.