Learn More
Strem, An Ascensus Company CAS# 863971-63-5. 100mg. Carbonylhydrido[6-(di-t-butylphosphinomethylene)-2-(N,N-diethylaminomethyl)-1,6-dihydropyridine]ruthenium(II), min. 98% Milstein Catalyst. MFCD11045093
SDP
Supplier:  Strem, An Ascensus Company 440091100MG
Encompass_Preferred
CAS# 863971-63-5. 100mg. Carbonylhydrido[6-(di-t-butylphosphinomethylene)-2-(N,N-diethylaminomethyl)-1,6-dihydropyridine]ruthenium(II), min. 98% Milstein Catalyst. MFCD11045093. Molecular Weight: 451.55. Molecular Formula: C20H35N2OPRu. Color/form: red-black solid Strem# 44-0091. http://www.strem.com/catalog/v/44-0091/
Catalog No. 50-901-11766
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.