Learn More
AdipoGen Carbophenothion
SDP
Supplier:  AdipoGen CDXC0683M500
Encompass_Preferred
Chemical. CAS 786-19-6. Formula C11H16ClO2PS3. MW 342.87. . An organophosphate pesticide. Insecticide and acaricide used in the agriculture industry. Acetylcholinesterase AChE inhibitor in the nervous system with subsequent accumulation of toxic levels of acetylcholine ACh. Used for citrus fruits against insects. Compound can be used as analytical reference material.
Catalog No. 50-256-1352
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.