Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Carboxine

Catalog No. AC467620250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467620250 25 g
AC467620010 1 g
AC467620050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC467620250 Supplier Thermo Scientific Chemicals Supplier No. 467620250
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5234-68-4 | C12H13NO2S, C12H13NO2S | 235.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5234-68-4
Molecular Formula C12H13NO2S

Specifications

Quantity 25 g
Melting Point 89.0°C to 95.0°C
Formula Weight 235.31
Chemical Name or Material Carboxine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause an allergic skin reaction.

May cause damage to organs through prolonged or repeated exposure if inhaled.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Take off all contaminated clothing and wash it before reuse Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation or rash occurs: Get medical advice/attention.

Get medical attention/advice if you feel unwell.

EINECSNumber 226-031-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.