Learn More
AdipoGen Carboxy-DCFDA
SDP
Supplier:  AdipoGen CDXC0042G001
Encompass_Preferred
Chemical. CAS 127770-45-0. Formula C25H14Cl2O9. MW 529.28. Synthetic. Intracellular esterase substrate with low sensitivity of fluorescence to intracellar pH.
Catalog No. 50-256-1523
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.