Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Carboxy-DCFDA N-succinimidyl Ester, Fluorogenic Esterase Substrate, 250 mg

Catalog No. 502705333
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5333 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5333 Supplier Abcam Supplier No. AB145286250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable, fluorogenic esterase substrate that hydrolyzes intracellularly to a green fluorophore.

  • Molecular weight: 626.35 g/mol
  • Purity: >96%
  • Properties: Cell-permeable, fluorogenic esterase substrate; becomes hydrolyzed intracellularly to green fluorophore DCFA (ab145439) and covalently bonds to proteins

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% DCFDA N-succinimidyl ester
Substrate Properties Chemical Substrate
Target Molecule Proteins
Substrate Esterase substrate
Content And Storage Store at -20°C.
Detection Location In-Cell Detection
Shipping Condition Ambient
Detection Method Fluorescence
Formulation 100% DCFDA N-succinimidyl ester
Quantity 250 mg
For Use With (Application) Sample Prep and Detection
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.