Learn More
Abcam Carboxy-DCFDA N-succinimidyl Ester, Fluorogenic Esterase Substrate, >96% Purity, 50 mg
Description
- Molecular Weight: 626.35 g/mol
- Purity: >96%
- Product Type: Cell-permeable, fluorogenic esterase substrate
- Mechanism: Becomes hydrolyzed intracellularly to green fluorophore DCFA (ab145439) and covalently bonds to proteins
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Proteins |
| Substrate Properties | Chemical Substrate |
| Substrate | Carboxy-DCFDA N-succinimidyl ester |
| Shipping Condition | Ambient |
| Buffer | 100% DCFDA N-succinimidyl ester |
| Content And Storage | Store at -20°C. |
| Detection Location | In-Cell Detection |
| Detection Method | Fluorescence |
| Formulation | 100% DCFDA N-succinimidyl ester |
| Quantity | 50 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.