Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Directional Cloning of Protein-Coding Sequences
- Choose from a variety of initial applications (e.g., bacterial protein, mammalian or cell-free protein expression) and then transfer the insert as required
- Efficient insert transfer means direct use of recombinant clones with minimal time for screening background colonies
- Adaptable to high-throughput formats for large screening projects
Order Info
Kit supplied with all components necesssary for 50 transfer reactions; individual vectors sold separately
Specifications
Specifications
| Content And Storage | -30°C to -10°C |
| Description | Carboxy Flexi Cloning System, Transfer |
| Format | System |
| For Use With (Equipment) | Carboxy Flexi™ Cloning System |
| For Use With (Application) | Transferring Protein Coding Regions From Native Or N Terminal Flexi Vectors To C Terminal Flexi Vectors |
| No. of Reactions | 50 Transfer Reactions |
| Product Type | Carboxy Flexi Cloning System, Transfer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?