Learn More
Cayman Chemical Carboxy-PTIO (potassium salt), 148819-94-7, 5 mg
SDP
Supplier:  Cayman Chemical 815405
Encompass_Preferred
Carboxy-PTIO is a NO scavenger. It reacts stoichiometrically with NO and can be used for EPR detection of NO.{1203,4117}
Catalog No. NC9697883
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.