Learn More
Apexbio Technology LLC Carboxy-PTIO, potassium salt 10mg
Supplier: Apexbio Technology LLC B644510
Carboxy-PTIO potassium salt (CAS 148819-94-7) is a small molecule recognized as a nitric oxide (NO) scavenger It reacts rapidly with NO converting it to nitrogen oxides and thereby reducing bioavailable NO levels In lipopolysaccharide-induced rat models administration of Carboxy-PTIO potassium salt has been shown to attenuate hypotension and decrease the incidence of endotoxic shock via NO clearance This compound is widely employed in studies investigating NO-related signaling pathways and the pathophysiological roles of aberrant NO production
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.