Learn More
Apexbio Technology LLC Aminoguanidine hydrochloride 1937-19-5 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC B64525.1
Aminoguanidine hydrochloride (CAS 1937-19-5) is a small molecule inhibitor targeting diamine oxidase and nitric oxide synthase It has been shown to attenuate doxorubicin-induced apoptosis in a dose-dependent manner in experimental models In addition to its enzyme inhibitory activities aminoguanidine hydrochloride exhibits antioxidant properties These characteristics make it a widely used tool in research focused on the pathophysiology of diabetic nephropathy and related oxidative stress-mediated conditions
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.