Learn More
Medchemexpress LLC Carboxyatractyloside dipotassium | 33286-30-5 | 99.6% | 847.00 g/mol | C31H44K2O18S2 | 10 MG

Supplier: Medchemexpress LLC HYN252210MG
Carboxyatractyloside dipotassium is the dipotassium salt of carboxyatractyloside, a mitochondrial ADP/ATP carrier inhibitor used as a research reagent to study mitochondrial function, calcium signaling, and oxidative stress.
- High purity - 99.6% by RP-HPLC.
- CAS number 33286-30-5 for unambiguous identification.
- Molecular weight 847.00 g/mol.
- White to off-white solid appearance.
- Recommended storage: -20°C (sealed); in solvent -80°C up to 6 months.
- Supplied as a 10 mg mass for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.