Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxymethylcellulose Calcium Salt, Spectrum™ Chemical
SDP

Catalog No. 18601466 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-466 25 g Poly Bottle
18-601-467 100 g Poly Bottle
18-601-468 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-466 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C152825GM
Add to Cart
Add to Cart

C1528, 9050-04-8

Spectrum™ Chemical Carboxymethylcellulose Calcium Salt , also known as Carboxymethyl cellulose or cellulose gum, can be used as a thickening agent in pharmaceuticals. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9050-04-8

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 25 g
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Carboxymethylcellulose Calcium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.