Learn More
Justrite™ Polypropylene, Delrin, ETFE, Carboy Adapter Fittings, Six 1/8 in. Fittings and Six 1/4 in. Plugs
GREENER_CHOICE
Catalog No. 19830508 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
VaporTrap solvent waste system adapter
Catalog No. For Use With (Equipment)
19-830-508 VaporTrap solvent waste system adapter
1 options
Catalog No. 19-830-508 Supplier Justrite™ Supplier No. 12851
Add to Cart
Add to Cart
For Use With: VaporTrap solvent waste system adapter
  • Includes nut, ferrule, fitting plugs, filter fitting, elbow fitting, filter fitting plug
Specifications
Description Carboy Adapter Fittings
For Use With (Equipment) VaporTrap solvent waste system adapter
Height (English) 1 in.
Material ETFE, Delrin, Polypropylene
Product Type Fitting
Width (English) 1 in.